1,1'-(1,6-Hexandiyl)bis(1H-pyrrol-2,5-dion) 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: BMH

H58874.MC H58874.ME
H58874.MCEA 142 EUR
H58874.MC H58874.ME
1,1'-(1,6-Hexandiyl)bis(1H-pyrrol-2,5-dion) 97%
BMH (1,6-Bis(maleimido)hexane)

Purity: 97% CAS No.: 4856-87-5 Molecular Formula: C14H16N2O4 Formula Weight: 276.29 Storage Temperature: 2°C to 8°C Melting Point: 144° MDL No.: MFCD00047122

Formel: C₁₄H₁₆N₂O₄
Molecular Weight: 276,29 g/mol
Schmelzpunkt: 144 °C
MDL: MFCD00047122
CAS-Nummer: 4856-87-5
EINECS: 225-449-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR