β-Carotin 99%
Lieferant: Thermo Scientific
Synonyme:
2,2'-((1E,3E,5E,7E,9E,11E,13E,15E,17E)-3,7,12,16-Tetramethyloctadeca-1,3,5,7,9,11,13,15,17-nonaene-1,18-diyl)bis(1,3,3-trimethylcyclohex-1-ene), Provitamin A
H60106.03
H60106.06
H60106.03EA
42.5
EUR
H60106.03
H60106.06
β-Carotin 99%
beta-Carotene
Purity: 99%
CAS No.: 7235-40-7
Molecular Formula: C40H56
Formula Weight: 536.87
Storage Temperature: -30°C to -10°C
Flash Point: 103°(217°F)
Physical Form: Solid
Density: 1.000
Sensitivity: Air and Light Sensitive
Melting Point: 178-179°
Beilstein Reference: 1917416
MDL No.: MFCD00001556
Formel:
C₄₀H₅₆ Molecular Weight: 536,89 g/mol Schmelzpunkt: 178…179 °C Dichte: 1,000 Flammpunkt: 103 °C (217 °F) |
MDL:
MFCD00001556 CAS-Nummer: 7235-40-7 EINECS: 230-636-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...