N-alpha-BOC-D-Asparagin 95%
Lieferant: Thermo Scientific
Synonyme:
Boc-D-Asn-OH, Nɑ-Boc-D-asparagine
H61121.06
H61121.14
H61121.06EA
44.5
EUR
H61121.06
H61121.14
N-alpha-BOC-D-Asparagin 95%
N-alpha-BOC-D-Asparagin
Purity: 95%
CAS No.: 75647-01-7
Molecular Formula: C9H16N2O5
Formula Weight: 232.23
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
Beilstein Reference: 4843040
Optical Rotation: +7.3 (c=2 in DMF)
MDL No.: MFCD00065558
Formel:
C₉H₁₆N₂O₅ Molecular Weight: 232,24 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00065558 CAS-Nummer: 75647-01-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...