Boc-D-homophenylalanine 98%

Lieferant: Thermo Scientific

Synonyme: Boc-D-Homophe-OH, (R)-2-(Boc-amino)-4-phenylbutyric acid, Boc-D-HoPhe-OH

H61183.03 H61183.06
H61183.03EA 57.5 EUR
H61183.03 H61183.06
Boc-D-homophenylalanine 98%
Boc-D-homophenylalanine

Purity: 98% CAS No.: 82732-07-8 Molecular Formula: C15H21NO4 Formula Weight: 279.33 Physical Form: Powder Appearance: White to off-white Melting Point: 71-78° Beilstein Reference: 3653505 MDL No.: MFCD00076905

Formel: C₁₅H₂₁NO₄
Molecular Weight: 279,336 g/mol
Schmelzpunkt: 71…78 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00076905
CAS-Nummer: 82732-07-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR