N-Boc-D-serine methyl ester 97%
Lieferant: Thermo Fisher Scientific
Synonyme:
N-(tert-Butoxycarbonyl)-D-serine methyll ester, Boc-D-Ser-OMe, N-Boc-D-serine methyll ester
H61296.06
H61296.14
H61296.06EA
79.4
EUR
H61296.06
H61296.14
N-Boc-D-serine methyl ester 97%
N-Boc-D-serine methyl ester
Purity: 97%
CAS No.: 95715-85-8
Molecular Formula: C9H17NO5
Formula Weight: 219.23
Flash Point: 113°(235°F)
Physical Form: Liquid
Density: 1.080
Refractive Index: 1.4530
Appearance: Colourless to yellow
Boiling Point: 215°
Optical Rotation: +18° (c=5 in methanol)
MDL No.: MFCD00270516
Formel:
C₉H₁₇NO₅ Siedepunkt: 215 °C Dichte: 1,080 Flammpunkt: 113 °C (235 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00270516 CAS-Nummer: 95715-85-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...