2-Amino-6-(trifluoromethoxy)benzothiazole 98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
6-(Trifluoromethoxy)benzo[d]thiazol-2-amine, Riluzole
H61328.06
H61328.14
H61328.06EA
110
EUR
H61328.06
H61328.14
2-Amino-6-(trifluoromethoxy)benzothiazole 98%
2-Amino-6-(trifluoromethoxy)benzothiazole
Purity: 98%
CAS No.: 1744-22-5
Molecular Formula: C8H5F3N2OS
Formula Weight: 234.20
Physical Form: Solid
Appearance: White to yellow
Melting Point: 114-118°
Beilstein Reference: 748759
UN No.: UN2811
MDL No.: MFCD00210213
Formel:
C₈H₅F₃N₂OS Molecular Weight: 234,20 g/mol Schmelzpunkt: 114…118 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00210213 CAS-Nummer: 1744-22-5 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...