4,6-Dichlornicotinsäure 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4,6-dichloropyridine-3-carboxylic acid
H61381.06
H61381.14
H61381.06EA
104
EUR
H61381.06
H61381.14
4,6-Dichlornicotinsäure 97%
4,6-Dichloronicotinic acid
Purity: 97%
CAS No.: 73027-79-9
Molecular Formula: C6H3Cl2NO2
Formula Weight: 192.00
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: White
MDL No.: MFCD00234146
Formel:
C₆H₃Cl₂NO₂ Molecular Weight: 192,00 g/mol |
MDL:
MFCD00234146 CAS-Nummer: 73027-79-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...