(2,6-dimethylphenyl)methanol 98%
Lieferant: Thermo Fisher Scientific
Synonyme:
2,6-Dimethylbenzyl alcohol
H61409.MD
H61409.03
H61409.06
H61409.MDEA
39
EUR
H61409.MD
H61409.03
H61409.06
(2,6-dimethylphenyl)methanol 98%
(2,6-dimethylphenyl)methanol
Purity: 98%
CAS No.: 62285-58-9
Molecular Formula: C9H12O
Formula Weight: 136.19
Physical Form: Solid
MDL No.: MFCD00128004
Formel:
C₉H₁₂O Molecular Weight: 136.194 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00128004 CAS-Nummer: 62285-58-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...