Thiophen-3-methylamin 97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
3-Thienylmethylamine
H61487.03
H61487.06
H61487.03EA
173
EUR
H61487.03
H61487.06
Thiophen-3-methylamin 97%
Thiophen-3-methylamin
Purity: 97%
CAS No.: 27757-86-4
Molecular Formula: C5H7NS
Formula Weight: 113.18
Storage Temperature: 2°C to 8°C
Refractive Index: 1.5660
Boiling Point: 102°/15mm
UN No.: UN2735
MDL No.: MFCD01529872
Formel:
C₅H₇NS Molecular Weight: 113,18 g/mol Siedepunkt: 102 °C (15 mmHg) Dichte: 1,130 |
MDL:
MFCD01529872 CAS-Nummer: 27757-86-4 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...