(R)-Methyl-2-((tert-butoxycarbonyl)amino)-3-iodopropanoate 98%
Lieferant: Thermo Scientific
Synonyme:
Boc-β-iodo-Ala-OMe, BOC-3-iodo-L-alanine methyl ester, N-Boc-3-iodo-L-alanine Methyl Ester, Boc-β-Iodo-Ala-OMe, Boc-Ala(3-I)-OMe
H61638.03
H61638.06
H61638.03EA
72.1
EUR
H61638.03
H61638.06
(R)-Methyl-2-((tert-butoxycarbonyl)amino)-3-iodopropanoate 98%
(R)-Methyl-2-((tert-butoxycarbonyl)amino)-3-iodopropanoate
Purity: 98%
CAS No.: 93267-04-0
Molecular Formula: C9H16INO4
Formula Weight: 329.13
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Sensitivity: Light Sensitive
Appearance: White
Melting Point: 50-52°
Beilstein Reference: 4422011
MDL No.: MFCD00216579
Formel:
C₉H₁₆INO₄ Molecular Weight: 329,13 g/mol Schmelzpunkt: 50…52 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00216579 CAS-Nummer: 93267-04-0 |