6-Chlor-2,3-pyridindiamin 95%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
2,3-Diamino-6-chloropyridine
H61716.03
H61716.06
H61716.03EA
67.7
EUR
H61716.03
H61716.06
6-Chlor-2,3-pyridindiamin 95%
6-Chloropyridine-2,3-diamine
Purity: 95%
CAS No.: 40851-95-4
Molecular Formula: C5H6ClN3
Formula Weight: 143.57
Storage Temperature: -30°C to -10°C
UN No.: UN2811
MDL No.: MFCD00209966
Formel:
C₅H₆ClN₃ Molecular Weight: 143,57 g/mol |
MDL:
MFCD00209966 CAS-Nummer: 40851-95-4 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...