3-Bromanthranilsäure 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Amino-3-bromobenzoic acid

H61925.03 H61925.06
H61925.03EA 55.5 EUR
H61925.03 H61925.06
3-Bromanthranilsäure 97%
3-Bromanthranilsäure

Purity: 97% CAS No.: 20776-51-6 Molecular Formula: C7H6BrNO2 Formula Weight: 216.03 Flash Point: 161°(322°F) Physical Form: Solid Appearance: White to pale yellow Melting Point: 174-178° Beilstein Reference: 2936928 MDL No.: MFCD03618453

Formel: C₇H₆BrNO₂
Molecular Weight: 216,03 g/mol
Schmelzpunkt: 174…178 °C
Flammpunkt: 161 °C (322 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD03618453
CAS-Nummer: 20776-51-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR