Boc-Orn-OH 95%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Nα-Boc-L-ornithine
H62014.06
H62014.14
H62014.06EA
105
EUR
H62014.06
H62014.14
Boc-Orn-OH 95%
Boc-Orn-OH
Purity: 95%
CAS No.: 21887-64-9
Molecular Formula: C10H20N2O4
Formula Weight: 232.28
Physical Form: Powder
Appearance: White
Melting Point: 207-208°
Beilstein Reference: 4313353
MDL No.: MFCD00076970
Formel:
C₁₀H₂₀N₂O₄ Molecular Weight: 232,28 g/mol Schmelzpunkt: 207…208 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00076970 CAS-Nummer: 21887-64-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...