2-Amino-3-fluorobenzeneboronic acid pinacol ester 96%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Fluoro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 2-Amino-3-fluorophenylboronic acid pinacol ester, Benzenamine, 2-fluoro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-
H62033.MD
H62033.03
H62033.MDEA
69.5
EUR
H62033.MD
H62033.03
2-Amino-3-fluorobenzeneboronic acid pinacol ester 96%
2-Amino-3-fluorobenzeneboronic acid pinacol ester
Purity: 96%
Molecular Formula: C12H17BFNO2
Formula Weight: 237.08
Physical Form: Solid
Formel:
C₁₂H₁₇BFNO₂ Molecular Weight: 237,08 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12025468 CAS-Nummer: 1310384-26-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...