2-Bromo-6-fluorobenzylamine hydrochloride 96%

Lieferant: Thermo Fisher Scientific

Synonyme: 1-(2-Bromo-6-fluorophenyl)methanamine hydrochloride (1:1), Benzenemethanamine, 2-bromo-6-fluoro-, hydrochloride (1:1)

H62086.03 H62086.06
H62086.03EA 209 EUR
H62086.03 H62086.06
2-Bromo-6-fluorobenzylamine hydrochloride 96%
2-Bromo-6-fluorobenzylamine hydrochloride

Purity: 96% CAS No.: 1214330-90-1 Molecular Formula: C7H7BrFN.HCl Formula Weight: 240.50 Physical Form: Solid

Formel: C₇H₇BrFN·HCl
Molecular Weight: 240.50 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12546112
CAS-Nummer: 1214330-90-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR