(S)-2,3-Diaminopropanoic acid hydrochloride 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: H-Dap-OH.HCl, H-Dap-OH·HCl, L-3-Aminoalanine Hydrochloride, (S)-2,3-Diaminopropionic acid monohydrochloride

H62128.03 H62128.06
H62128.03EA 96 EUR
H62128.03 H62128.06
(S)-2,3-Diaminopropanoic acid hydrochloride 95%
(S)-2,3-Diaminopropanoic acid hydrochloride

Purity: 95% CAS No.: 1482-97-9 Molecular Formula: C3H8N2O2.HCl Formula Weight: 140.57 Storage Temperature: -30°C to -10°C Physical Form: Crystalline solid Sensitivity: Hygroscopic Appearance: Off-white Melting Point: 231-233° dec Beilstein Reference: 4263944 Optical Rotation: +24°(c=2 in 0.5 M HCl) MDL No.: MFCD00065497

Formel: C₃H₈N₂O₂·HCl
Molecular Weight: 140,6 g/mol
Schmelzpunkt: 231…233 °C (Zers.)
Lagertemperatur: Gefrierschrank
MDL: MFCD00065497
CAS-Nummer: 1482-97-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR