2-Bromo-4-fluorobenzylamine hydrochloride 96%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
(2-Bromo-4-fluorophenyl)methanamine hydrochloride
H62200.03
H62200.06
H62200.03EA
209
EUR
H62200.03
H62200.06
2-Bromo-4-fluorobenzylamine hydrochloride 96%
2-Bromo-4-fluorobenzylamine hydrochloride
Purity: 96%
CAS No.: 289038-14-8
Molecular Formula: C7H7BrFN.HCl
Formula Weight: 240.50
Physical Form: Solid
MDL No.: MFCD00672922
Formel:
C₇H₈BrClFN Molecular Weight: 240.5 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00672922 CAS-Nummer: 289038-14-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...