Boc-D-Asp-OBzl 98%

Lieferant: Thermo Fisher Scientific

Synonyme: N-Boc-D-aspartic acid 1-benzyl ester

H62210.03 H62210.06
H62210.03EA 88.4 EUR
H62210.03 H62210.06
Boc-D-Asp-OBzl 98%
Boc-D-Asp-OBzl

Purity: 98% CAS No.: 92828-64-3 Molecular Formula: C16H21NO6 Formula Weight: 323.34 Physical Form: Powder Appearance: White MDL No.: MFCD00038264

Formel: C₁₆H₂₁NO₆
Lagertemperatur: Raumtemperatur
MDL: MFCD00038264
CAS-Nummer: 92828-64-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR