Boc-Arg(NO₂)-OH 98%
Lieferant: Thermo Scientific
Synonyme:
Nα-Boc-Nomega-nitro-L-arginine
H62339.14
H62339.22
H62339.14EA
88.4
EUR
H62339.14
H62339.22
Boc-Arg(NO₂)-OH 98%
Boc-Arg(NO₂)-OH
Purity: 98%
CAS No.: 2188-18-3
Molecular Formula: C11H21N5O6
Formula Weight: 319.31
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Beilstein Reference: 2015163
MDL No.: MFCD00065556
Formel:
C₁₁H₂₁N₅O₆ Molecular Weight: 319,318 g/mol |
MDL:
MFCD00065556 CAS-Nummer: 2188-18-3 EINECS: 218-580-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...