(S)-4-Amino-2-(tert-butoxycarbonylamino)butyric acid 97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: Boc-Dab-OH, (S)-4-Amino-2-(Boc-amino)butyric Acid, (S)-4-Amino-2-(tert-butoxycarbonylamino)butyric Acid, (S)-Nalpha-Boc-2,4-diaminobutyric Acid, (S)-4-Amino-2-(Boc-amino)butyric acid (S)-Nα-Boc-2,4-diaminobutyric acid Boc-Dab-OH

H62450.03 H62450.06
H62450.03EA 44.1 EUR
H62450.03 H62450.06
(S)-4-Amino-2-(tert-butoxycarbonylamino)butyric acid 97%
(S)-4-Amino-2-(tert-butoxycarbonylamino)butyric acid

Purity: 97% CAS No.: 25691-37-6 Molecular Formula: C9H18N2O4 Formula Weight: 218.25 Physical Form: Powder Appearance: White to off-white Beilstein Reference: 4745487 Optical Rotation: +7°(c=1 in methanol) MDL No.: MFCD00236841

Formel: C₉H₁₈N₂O₄
Molecular Weight: 218,25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00236841
CAS-Nummer: 25691-37-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR