Methyl-4-bromo-2-fluorophenylacetate 96%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Methyl 2-(4-bromo-2-fluorophenyl)acetate
H62482.MD
H62482.03
H62482.MDEA
73.9
EUR
H62482.MD
H62482.03
Methyl-4-bromo-2-fluorophenylacetate 96%
Methyl-4-bromo-2-fluorophenylacetate
Purity: 96%
CAS No.: 193290-19-6
Molecular Formula: C9H8BrFO2
Formula Weight: 247.06
Physical Form: Solid
Formel:
C₉H₈BrFO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD20483545 CAS-Nummer: 193290-19-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...