L-(-)-Tryptophanol 97%

Lieferant: Thermo Scientific

Synonyme: L-Tryptophanol, (S)-(-)-2-Amino-3-(3-indolyl)propanol, (S)-2-Amino-3-(1H-indol-3-yl)propan-1-ol

H62506.03 H62506.06
H62506.03EA 113 EUR
H62506.03 H62506.06
L-(-)-Tryptophanol 97%
L-(-)-Tryptophanol

Purity: 97% CAS No.: 2899-29-8 Molecular Formula: C11H14N2O Formula Weight: 190.24 Physical Form: Viscous liquid Appearance: Brown Optical Rotation: -20° (c=1 in methanol) MDL No.: MFCD00037970

Formel: C₁₁H₁₄N₂O
Molecular Weight: 190,25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00037970
CAS-Nummer: 2899-29-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR