trans-4-Cyanocyclohexylamine hydrochloride 97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
(1r,4r)-4-aminocyclohexane-1-carbonitrile hydrochloride, trans-4-Aminocyclohexanecarbonitrile hydrochloride
H62522.MD
H62522.03
H62522.MDEA
161
EUR
H62522.MD
H62522.03
trans-4-Cyanocyclohexylamine hydrochloride 97%
trans-4-Cyanocyclohexylamine hydrochloride
Purity: 97%
CAS No.: 873537-33-8
Molecular Formula: C7H12N2.HCl
Formula Weight: 160.64
UN No.: UN3439
MDL No.: MFCD21643021
Formel:
C₇H₁₂N₂·ClH Molecular Weight: 160,645 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD21643021 CAS-Nummer: 873537-33-8 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...