3-Fluoro-4-(4-morpholinylmethyl)benzeneboronic acid pinacol ester 96%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
4-(2-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)morpholine
H62530.MD
H62530.03
H62530.MDEA
115
EUR
H62530.MD
H62530.03
3-Fluoro-4-(4-morpholinylmethyl)benzeneboronic acid pinacol ester 96%
3-Fluoro-4-(4-morpholinylmethyl)benzeneboronic acid pinacol ester
Purity: 96%
CAS No.: 1073354-74-1
Molecular Formula: C17H25BFNO3
Formula Weight: 321.19
Physical Form: Solid
Melting Point: 35-40°
MDL No.: MFCD09027076
Formel:
C₁₇H₂₅BFNO₃ Schmelzpunkt: 35…40 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09027076 CAS-Nummer: 1073354-74-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...