Boc-His-OMe 95%

Lieferant: Thermo Scientific

Synonyme: N-Boc-L-histidine methyl ester

H62545.03 H62545.06
H62545.03EA 87.4 EUR
H62545.03 H62545.06
Boc-His-OMe 95%
Boc-His-OMe

Purity: 95% CAS No.: 2488-14-4 Molecular Formula: C12H19N3O4 Formula Weight: 269.30 Physical Form: Solid MDL No.: MFCD00167635

Formel: C₁₂H₁₉N₃O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00167635
CAS-Nummer: 2488-14-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR