Boc-Asp-OMe 98%

Lieferant: Thermo Fisher Scientific

Synonyme: Boc-Asp-OMe, N-Boc-L-aspartic acid 1-Methyl ester, N-Boc-L-aspartic acid 1-methyl ester, 1-Methyl N-Boc-L-aspartate, 1-Methyl N-(tert-Butoxycarbonyl)-L-aspartate, N-(tert-Butoxycarbonyl)-L-aspartic acid 1-Methyl ester

H62590.03 H62590.06
H62590.03EA 108 EUR
H62590.03 H62590.06
Boc-Asp-OMe 98%
Boc-Asp-OMe

Purity: 98% CAS No.: 98045-03-5 Molecular Formula: C10H17NO6 Formula Weight: 247.25 Physical Form: Solid MDL No.: MFCD03094777

Formel: C₁₀H₁₇NO₆
Lagertemperatur: Raumtemperatur
MDL: MFCD03094777
CAS-Nummer: 98045-03-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR