Boc-Ala-NH2 96%

Lieferant: Thermo Scientific

Synonyme: tert-butyl N-(1-carbamoylethyl)carbamate, Boc-Ala-NH₂, N-Boc-L-alaninamide

H62604.03 H62604.06
H62604.03EA 183 EUR
H62604.03 H62604.06
Boc-Ala-NH2 96%
Boc-Ala-NH2

Purity: 96% CAS No.: 85642-13-3 Molecular Formula: C8H16N2O3 Formula Weight: 188.22 Physical Form: Solid MDL No.: MFCD09263333

Formel: C₈H₁₆N₂O₃
Molecular Weight: 188,227 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03701447
CAS-Nummer: 85642-13-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR