Ethyl-N-{[(2-methyl-2-propanyl)oxy]carbonyl}glycinat 95%
Lieferant: Thermo Fisher Scientific
Synonyme:
N-(tert-Butoxycarbonyl)glycine ethyl ester, ethyl 2-((tert-Butoxycarbonyl)amino)acetate, Boc-Gly-OEt, N-Boc-glycine ethyl ester
H62611.06
H62611.14
H62611.06EA
57.8
EUR
H62611.06
H62611.14
Ethyl-N-{[(2-methyl-2-propanyl)oxy]carbonyl}glycinat 95%
ethyl-2-((tert-butoxycarbonyl)amino)acetate
Purity: 95%
CAS No.: 14719-37-0
Molecular Formula: C9H17NO4
Formula Weight: 203.24
Physical Form: Oily liquid
Appearance: Colourless
MDL No.: MFCD05663720
Formel:
C₉H₁₇NO₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD05663720 CAS-Nummer: 14719-37-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...