(S)-3-(Benzyloxy)-2-((tert-butoxycarbonyl)amino)propanoic acid 97%
Lieferant: Thermo Fisher Scientific
Synonyme:
Boc-Ser(Bzl)-OH, N-Boc-O-benzyl-L-serine
H62705.06
H62705.14
H62705.06EA
47.3
EUR
H62705.06
H62705.14
(S)-3-(Benzyloxy)-2-((tert-butoxycarbonyl)amino)propanoic acid 97%
(S)-3-(Benzyloxy)-2-((tert-butoxycarbonyl)amino)propanoic acid
Purity: 97%
CAS No.: 23680-31-1
Molecular Formula: C15H21NO5
Formula Weight: 295.33
Physical Form: Solid
Melting Point: 58-60°
Beilstein Reference: 3064461
Optical Rotation: +20°(c=2 in ethanol)
MDL No.: MFCD00066063
Formel:
C₁₅H₂₁NO₅ Molecular Weight: 295.3 g/mol Schmelzpunkt: 58…60 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00066063 CAS-Nummer: 23680-31-1 EINECS: 245-820-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...