Boc-Thr(tBu)-OH 95%
Lieferant: Thermo Scientific
Synonyme:
(2S,3R)-3-(tert-Butoxy)-2-((tert-butoxycarbonyl)amino)butanoic acid, N-Boc-O-tert-butyl-L-threonine
H62713.03
H62713.06
H62713.03EA
45.6
EUR
H62713.03
H62713.06
Boc-Thr(tBu)-OH 95%
Boc-Thr(tBu)-OH
Purity: 95%
CAS No.: 13734-40-2
Molecular Formula: C13H25NO5
Formula Weight: 275.34
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Melting Point: 95-98°
Beilstein Reference: 4454820
Optical Rotation: +3°(c=2 in DMF)
MDL No.: MFCD00151115
Formel:
C₁₃H₂₅NO₅ Molecular Weight: 275,345 g/mol Schmelzpunkt: 95…98 °C |
MDL:
MFCD00151115 CAS-Nummer: 13734-40-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...