2-Fluoro-6-methoxybenzeneboronic acid pinacol ester 96%

Lieferant: Thermo Scientific

Synonyme: 1,3,2-Dioxaborolane, 2-(2-fluoro-6-methoxyphenyl)-4,4,5,5-tetramethyl-, 2-Fluoro-6-methoxyphenylboronic acid pinacol ester, 2-(2-Fluoro-6-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

H62812.MD H62812.03
H62812.MDEA 124 EUR
H62812.MD H62812.03
2-Fluoro-6-methoxybenzeneboronic acid pinacol ester 96%
2-Fluoro-6-methoxybenzeneboronic acid pinacol ester

Purity: 96% Molecular Formula: C13H18BFO3 Formula Weight: 252.09 Physical Form: Solid

Formel: C₁₃H₁₈BFO₃
Molecular Weight: 252,09 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16996294
CAS-Nummer: 1599432-41-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR