2-Carbamoyl-3-fluorobenzeneboronic acid pinacol ester 96%

Lieferant: Thermo Scientific

Synonyme: 2-Carbamoyl-3-fluorophenylboronic acid pinacol ester

H62904.MD H62904.03
H62904.MDEA 157 EUR
H62904.MD H62904.03
2-Carbamoyl-3-fluorobenzeneboronic acid pinacol ester 96%
2-Carbamoyl-3-fluorobenzeneboronic acid pinacol ester

Purity: 96% Molecular Formula: C13H17BFNO3 Formula Weight: 265.09 Physical Form: Solid

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR