Fmoc-D-Orn(Boc)-OH 95%

Lieferant: Thermo Scientific

Synonyme: Nδ-Boc-Nα-Fmoc-D-ornithine

H62930.03 H62930.06
H62930.03EA 62.7 EUR
H62930.03 H62930.06
Fmoc-D-Orn(Boc)-OH 95%
Fmoc-D-Orn(Boc)-OH

Purity: 95% CAS No.: 118476-89-4 Molecular Formula: C25H30N2O6 Formula Weight: 454.52 Physical Form: Powder Appearance: Yellow MDL No.: MFCD00077065

Formel: C₂₅H₃₀N₂O₆
Lagertemperatur: Raumtemperatur
MDL: MFCD00077065
CAS-Nummer: 118476-89-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR