Fmoc-D-Dab(Boc)-OH 98%

Lieferant: Thermo Fisher Scientific

Synonyme: (R)-4-(Boc-amino)-2-(Fmoc-amino)butyric acid

H62954.03 H62954.06
H62954.03EA 144 EUR
H62954.03 H62954.06
Fmoc-D-Dab(Boc)-OH 98%
Fmoc-D-Dab(Boc)-OH

Purity: 98% CAS No.: 114360-56-4 Molecular Formula: C24H28N2O6 Formula Weight: 440.49 Physical Form: Powder Appearance: White MDL No.: MFCD00798647

Formel: C₂₄H₂₈N₂O₆
Molecular Weight: 440.496 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00798647
CAS-Nummer: 114360-56-4

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR