H-Lys(boc)-OtBu·HCl 95%

Lieferant: Thermo Scientific

Synonyme: Nepsilon-Boc-L-lysine tert-butyl ester hydrochloride, H-Lys(Boc)-OtBu.HCl, H-Lys(Boc)-OtBu·HCl

H62993.03 H62993.06
H62993.03EA 114 EUR
H62993.03 H62993.06
H-Lys(boc)-OtBu·HCl 95%
H-Lys(boc)-OtBu·HCl

Purity: 95% CAS No.: 13288-57-8 Molecular Formula: C15H30N2O4.HCl Formula Weight: 338.87 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: White MDL No.: MFCD00038899

Formel: C₁₅H₃₁ClN₂O₄
Molecular Weight: 338,873 g/mol
MDL: MFCD00038899
CAS-Nummer: 13288-57-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR