3-phenylimidazolidine-2,4-dione 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3-Phenylimidazolidine-2,4-dione, 3-Phenylhydantoin

H63009.MD H63009.03
H63009.MDEA 51.4 EUR
H63009.MD H63009.03
3-phenylimidazolidine-2,4-dione 95%
3-phenylimidazolidine-2,4-dione

Purity: 95% CAS No.: 2221-13-8 Molecular Formula: C9H8N2O2 Formula Weight: 176.17 Physical Form: Solid Appearance: White MDL No.: MFCD00209486

Formel: C₉H₈N₂O₂
Molecular Weight: 176,175 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00209486
CAS-Nummer: 2221-13-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR