2-Fluor-6-nitrobenzoesäure 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 6-Fluoro-2-nitrobenzoic acid

H63013.06 H63013.14 H63013.22
H63013.06EA 52.9 EUR
H63013.06 H63013.14 H63013.22
2-Fluor-6-nitrobenzoesäure 98%
2-Fluoro-6-nitrobenzoic acid

Purity: 98% CAS No.: 385-02-4 Molecular Formula: C7H4FNO4 Formula Weight: 185.11 Physical Form: Solid MDL No.: MFCD01862079

Formel: C₇H₄FNO₄
Molecular Weight: 185,11 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01862079
CAS-Nummer: 385-02-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR