S-Trityl-D-cysteine 98%
Lieferant: Thermo Scientific
Synonyme:
(S)-2-Amino-3-(tritylthio)propanoic acid, H-D-Cys(Trt)-OH
H63022.03
H63022.06
H63022.03EA
42
EUR
H63022.03
H63022.06
S-Trityl-D-cysteine 98%
S-Trityl-D-cysteine
Purity: 98%
CAS No.: 25840-82-8
Molecular Formula: C22H21NO2S
Formula Weight: 363.47
Storage Temperature: 2°C to 8°C
Physical Form: Solid
MDL No.: MFCD00236948
Formel:
C₂₂H₂₁NO₂S Molecular Weight: 363,475 g/mol |
MDL:
MFCD00236948 CAS-Nummer: 25840-82-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...