Methyl-3-amino-5-fluorobenzoate 98%
Lieferant: Thermo Scientific
Synonyme: Methyl 3-amino-5-fluorobenzoate
H63038.03 H63038.06
H63038.03EA 87.3 EUR
H63038.03 H63038.06
Methyl-3-amino-5-fluorobenzoate 98%
Methyl-3-amino-5-fluorobenzoate
Purity: 98% CAS No.: 884497-46-5 Molecular Formula: C8H8FNO2 Formula Weight: 169.16 Physical Form: Solid Appearance: White to yellow MDL No.: MFCD05863683
Formel: C₈H₈FNO₂
Molecular Weight: 169,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05863683
CAS-Nummer: 884497-46-5