Fmoc-O-methyl-D-tyrosine 98%

Lieferant: Thermo Fisher Scientific

Synonyme: N-Fmoc-O-methyl-D-tyrosine, Fmoc-D-Tyr(Me)-OH

H63073.03 H63073.06
H63073.03EA 48.2 EUR
H63073.03 H63073.06
Fmoc-O-methyl-D-tyrosine 98%
Fmoc-O-methyl-D-tyrosine

Purity: 98% CAS No.: 201335-88-8 Molecular Formula: C25H23NO5 Formula Weight: 417.45 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White MDL No.: MFCD00237393

Formel: C₂₅H₂₃NO₅
Molecular Weight: 417,461 g/mol
MDL: MFCD00237393
CAS-Nummer: 201335-88-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR