1-Boc-4-(aminomethyl)-4-hydroxypiperidine 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
tert-Butyl 4-(aminomethyl)-4-hydroxypiperidine-1-carboxylate, 1-Boc-4-(Aminomethyl)-4-hydroxypiperidine
H63079.03
H63079.06
H63079.03EA
88.9
EUR
H63079.03
H63079.06
1-Boc-4-(aminomethyl)-4-hydroxypiperidine 97%
1-Boc-4-(aminomethyl)-4-hydroxypiperidine
Purity: 97%
CAS No.: 392331-66-7
Molecular Formula: C11H22N2O3
Formula Weight: 230.3
Physical Form: Solid
Sensitivity: Air Sensitive
Appearance: Off-white
MDL No.: MFCD11579803
Formel:
C₁₁H₂₂N₂O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD11579803 CAS-Nummer: 392331-66-7 |