O-Methyl-L-tyrosine 98%
Lieferant: Thermo Fisher Scientific
Synonyme:
(S)-2-Amino-3-(4-methoxyphenyl)propionic Acid, H-Tyr(Me)-OH, 4-Methoxy-L-phenylalanine
H63096.03
H63096.06
H63096.03EA
43.1
EUR
H63096.03
H63096.06
O-Methyl-L-tyrosine 98%
O-Methyl-L-tyrosine
Purity: 98%
CAS No.: 6230-11-1
Molecular Formula: C10H13NO3
Formula Weight: 195.22
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Melting Point: 259-261° dec
Beilstein Reference: 2212726
Optical Rotation: -7°(c=1 in 1M HCl)
MDL No.: MFCD00002604
Formel:
C₁₀H₁₃NO₃ Molecular Weight: 195.22 g/mol Schmelzpunkt: 259…261 °C (Zers.) |
MDL:
MFCD00002604 CAS-Nummer: 6230-11-1 EINECS: 228-333-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...