Methyl-3-cyanoindole-6-carboxylate 97%
Lieferant: Thermo Scientific
Achtung
Synonyme: Methyl 3-cyano-1H-indole-6-carboxylate
H63100.MD H63100.03 H63100.06
H63100.MDEA 71.5 EUR
H63100.MD H63100.03 H63100.06
Methyl-3-cyanoindole-6-carboxylate 97%
Methyl-3-cyanoindole-6-carboxylate
Purity: 97% CAS No.: 1000576-51-1 Molecular Formula: C11H8N2O2 Formula Weight: 200.19 Physical Form: Solid MDL No.: MFCD09878564
Formel: C₁₁H₈N₂O₂
Molecular Weight: 200,197 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09878564
CAS-Nummer: 1000576-51-1