Methyl-2-bromo-4-methylbenzoate 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Methyl 2-bromo-4-methylbenzoate
H63144.03
H63144.06
H63144.03EA
98.8
EUR
H63144.03
H63144.06
Methyl-2-bromo-4-methylbenzoate 98%
Methyl-2-bromo-4-methylbenzoate
Purity: 98%
CAS No.: 87808-49-9
Molecular Formula: C9H9BrO2
Formula Weight: 229.07
Storage Temperature: -30°C to -10°C
Physical Form: Solid
MDL No.: MFCD03840737
Formel:
C₉H₉BrO₂ Molecular Weight: 229.07 g/mol |
MDL:
MFCD03840737 CAS-Nummer: 87808-49-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...