Boc-Phe-OH 95%

Lieferant: Thermo Scientific

Synonyme: N-Boc-DL-phenylalanine, 2-((tert-Butoxycarbonyl)amino)-3-phenylpropanoic acid, Boc-DL-Phe-OH

H63164.03 H63164.06 H63164.14
H63164.03EA 30.4 EUR
H63164.03 H63164.06 H63164.14
Boc-Phe-OH 95%
Boc-Phe-OH

Purity: 95% CAS No.: 4530-18-1 Molecular Formula: C14H19NO4 Formula Weight: 265.3 Storage Temperature: 2°C to 8°C Physical Form: Solid MDL No.: MFCD00558969

Formel: C₁₄H₁₉NO₄
Molecular Weight: 265,309 g/mol
MDL: MFCD00558969
CAS-Nummer: 4530-18-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR