Methyl-6-bromo-1H-indole-2-carboxylate 97%
Lieferant: Thermo Fisher Scientific
H63187.03
H63187.06
H63187.03EA
85.1
EUR
H63187.03
H63187.06
Methyl-6-bromo-1H-indole-2-carboxylate 97%
Methyl-6-bromo-1H-indole-2-carboxylate
Purity: 97%
CAS No.: 103858-53-3
Molecular Formula: C10H8BrNO2
Formula Weight: 254.08
Storage Temperature: 2°C to 8°C
Physical Form: Crystalline powder
Appearance: White
MDL No.: MFCD03094942
Formel:
C₁₀H₈BrNO₂ |
MDL:
MFCD00590200 CAS-Nummer: 372089-59-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...