tert-Butyl-4-(4-aminophenyl)piperazine-1-carboxylate 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1-Boc-4-(4-aminophenyl)piperazine, tert-Butyl 4-(4-aminophenyl)piperazine-1-carboxylate
H63191.03
H63191.06
H63191.03EA
102
EUR
H63191.03
H63191.06
tert-Butyl-4-(4-aminophenyl)piperazine-1-carboxylate 97%
tert-Butyl-4-(4-aminophenyl)piperazine-1-carboxylate
Purity: 97%
CAS No.: 170911-92-9
Molecular Formula: C15H23N3O2
Formula Weight: 277.36
Physical Form: Solid
MDL No.: MFCD04115065
Formel:
C₁₅H₂₃N₃O₂ Molecular Weight: 277,36 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115065 CAS-Nummer: 170911-92-9 |