4-Ethyl-1H-pyrazol-3-amine 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3-Amino-4-ethyl-1H-pyrazole

H63199.03 H63199.06
H63199.03EA 194 EUR
H63199.03 H63199.06
4-Ethyl-1H-pyrazol-3-amine 98%
4-Ethyl-1H-pyrazol-3-amine

Purity: 98% CAS No.: 43024-15-3 Molecular Formula: C5H9N3 Formula Weight: 111.15 Physical Form: Solid MDL No.: MFCD06797570

Formel: C₅H₉N₃
Molecular Weight: 111,148 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06797570
CAS-Nummer: 43024-15-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR