2-Amino-3-(1H-indol-3-yl)propanamide 98%

Lieferant: Thermo Scientific

Synonyme: DL-Tryptophanamide

H63227.MD H63227.03
H63227.MDEA 47.7 EUR
H63227.MD H63227.03
2-Amino-3-(1H-indol-3-yl)propanamide 98%
2-Amino-3-(1H-indol-3-yl)propanamide

Purity: 98% CAS No.: 6720-02-1 Molecular Formula: C11H13N3O Formula Weight: 203.24 Physical Form: Crystalline powder Appearance: Off-white MDL No.: MFCD03826622

Formel: C₁₁H₁₃N₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD03826622
CAS-Nummer: 6720-02-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR