2-Fluor-3-nitrobenzoesäure 97%

Lieferant: Thermo Fisher Scientific
Achtung

H63235.03 H63235.06
H63235.03EA 44.3 EUR
H63235.03 H63235.06
2-Fluor-3-nitrobenzoesäure 97%
2-Fluoro-3-nitrobenzoic acid

Purity: 97% CAS No.: 317-46-4 Molecular Formula: C7H4FNO4 Formula Weight: 185.11 Storage Temperature: 2°C to 8°C Physical Form: Solid MDL No.: MFCD06797825

Formel: C₇H₄FNO₄
Molecular Weight: 185.11 g/mol
MDL: MFCD06797825
CAS-Nummer: 317-46-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR